Poly(tetrafluoroethylene)
Poly(tetrafluoroethylene)
Poly(tetrafluoroethylene)
Poly(tetrafluoroethylene)
Thermo Scientific Chemicals

Poly(tetrafluoroethylene)

CAS: 9002-84-0 | (C2F4)n | NaN g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
250 g
Packaging:
View all
Plastic bottle
Catalog number 178932500
Price (USD)
-
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS9002-84-0
Molecular Formula(C2F4)n
SMILESFC(F)(-*)C(F)(F)-*
MDL NumberMFCD00084455
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Appearance (Form)Powder
Loss on drying=<1 %

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only