p-Dithiane-2,5-diol, 96%
p-Dithiane-2,5-diol, 96%
p-Dithiane-2,5-diol, 96%
Thermo Scientific Chemicals

p-Dithiane-2,5-diol, 96%

CAS: 40018-26-6 | C4H8O2S2 | 152.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 179932500
Price (USD)
150.00
Each
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
150.00
Each
Chemical Identifiers
CAS40018-26-6
IUPAC Name1,4-dithiane-2,5-diol
Molecular FormulaC4H8O2S2
InChI KeyYUIOPHXTILULQC-UHFFFAOYNA-N
SMILESOC1CSC(O)CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Infrared spectrumConforms
GC>=95.0 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only