DL-2-Amino-3-methyl-1-butanol, 97%, Thermo Scientific Chemicals
DL-2-Amino-3-methyl-1-butanol, 97%, Thermo Scientific Chemicals
DL-2-Amino-3-methyl-1-butanol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

DL-2-Amino-3-methyl-1-butanol, 97%, Thermo Scientific Chemicals

CAS: 16369-05-4 | Molecular Formula: C5H13NO | Molecular Weight: 103.165 g/mol
Have Questions?
Catalog NumberQuantity
1804200505 g
Catalog number 180420050
Price (EUR)
76,30
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
76,30
Each
Chemical Identifiers
CAS16369-05-4
IUPAC Name(2S)-1-hydroxy-3-methylbutan-2-aminium
Molecular FormulaC5H14NO
InChI KeyNWYYWIJOWOLJNR-RXMQYKEDSA-O
SMILESCC(C)[C@H]([NH3+])CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4530 to 1.4570 (20°C, 589 nm)
Appearance (Color)Clear yellow
GC>=96.0 %

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only