3-Bromobenzoic acid, 99%
3-Bromobenzoic acid, 99%
3-Bromobenzoic acid, 99%
Thermo Scientific Chemicals

3-Bromobenzoic acid, 99%

CAS: 585-76-2 | C7H5BrO2 | 201.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number 181350250
Price (EUR)
38,65
Online Exclusive
42,50
Save 3,85 (9%)
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
38,65
Online Exclusive
42,50
Save 3,85 (9%)
Each
Chemical Identifiers
CAS585-76-2
IUPAC Name3-bromobenzoic acid
Molecular FormulaC7H5BrO2
InChI KeyVOIZNVUXCQLQHS-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Infrared spectrumConforms
Melting point154°C to 158°C
Appearance (Form)Powder
HPLC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only