Isopropanol, 99.5%, for analysis
Isopropanol, 99.5%, for analysis
Isopropanol, 99.5%, for analysis
Thermo Scientific Chemicals

Isopropanol, 99.5%, for analysis

CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
10 L
25 L
Packaging:
View all
EcoSafPak™ Glass Bottle
Catalog number 184130010
Price (USD)
68.65
Online exclusive
76.30
Save 7.65
Each
Quantity:
1 L
Packaging:
EcoSafPak™ Glass Bottle
Request bulk or custom format
Price (USD)
68.65
Online exclusive
76.30
Save 7.65
Each
Chemical Identifiers
CAS67-63-0
IUPAC Namepropan-2-ol
Molecular FormulaC3H8O
InChI KeyKFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILESCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Water=<0.1 % (Coulometric)
Residue after evaporation=<0.001 %
Copper (Cu)=<0.05 ppm
Magnesium (Mg)=<0.1 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only