1,3-Benzenedimethanol, 98%
1,3-Benzenedimethanol, 98%
1,3-Benzenedimethanol, 98%
Thermo Scientific Chemicals

1,3-Benzenedimethanol, 98%

CAS: 626-18-6 | C8H10O2 | 138.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Packaging:
Glass bottle
Catalog number 188200020
Price (EUR)
35,80
Each
Quantity:
2 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
35,80
Each
Chemical Identifiers
CAS626-18-6
IUPAC Name[3-(hydroxymethyl)phenyl]methanol
Molecular FormulaC8H10O2
InChI KeyYWMLORGQOFONNT-UHFFFAOYSA-N
SMILESOCC1=CC(CO)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Powder and/or chunks
Infrared spectrumConforms
Melting point54°C to 59°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only