Janus Green B, pure, high purity biological stain
Janus Green B, pure, high purity biological stain
Janus Green B, pure, high purity biological stain
Thermo Scientific Chemicals

Janus Green B, pure, high purity biological stain

CAS: 2869-83-2 | C30H31ClN6 | 511.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 191680050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2869-83-2
IUPAC Name3-(diethylamino)-7-{2-[4-(dimethylamino)phenyl]diazen-1-yl}-5-phenyl-5λ⁵-phenazin-5-ylium chloride
Molecular FormulaC30H31ClN6
InChI KeyXXACTDWGHQXLGW-UHFFFAOYSA-M
SMILES[Cl-].CCN(CC)C1=CC=C2N=C3C=CC(=CC3=[N+](C3=CC=CC=C3)C2=C1)N=NC1=CC=C(C=C1)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark green to dark brown to black
Infrared spectrumConforms
Appearance (Form)Powder
Dye content>=50 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only