Poly(ethylene glycol), average M.W. 1500
Poly(ethylene glycol), average M.W. 1500
Poly(ethylene glycol), average M.W. 1500
Thermo Scientific Chemicals

Poly(ethylene glycol), average M.W. 1500

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Quantity:
100 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 192261000
Price (JPY)
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS25322-68-3
Molecular Formula(C2H4O)n
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES[H]OCCO
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Hydroxyl value70 to 80 mg KOH/g
Appearance (Form)Waxy flakes
Water=<1.0 % (K.F.)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only