Sodium tetrachloropalladate(II), approx. 36% Pd
Sodium tetrachloropalladate(II), approx. 36% Pd
Sodium tetrachloropalladate(II), approx. 36% Pd
Thermo Scientific Chemicals

Sodium tetrachloropalladate(II), approx. 36% Pd

CAS: 13820-53-6 ∣ Cl4Na2Pd ∣ 294.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 195130010
Price (EUR)
124,00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
124,00
Each
Chemical Identifiers
CAS13820-53-6
IUPAC Namedisodium tetrachloropalladiumbis(ylium)
Molecular FormulaCl4Na2Pd
InChI KeyABKQFSYGIHQQLS-UHFFFAOYSA-J
SMILES[Na+].[Na+].Cl[Pd++](Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Silver (Ag)=<100 ppm
Copper (Cu)=<50 ppm
Iron (Fe)=<50 ppm
Magnesium (Mg)=<50 ppm
Silicon (Si)=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only