Iodine, 99.5%, extra pure, resublimed
Iodine, 99.5%, extra pure, resublimed
Iodine, 99.5%, extra pure, resublimed
Thermo Scientific Chemicals

Iodine, 99.5%, extra pure, resublimed

CAS: 7553-56-2 | I2 | 253.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 196561000
Price (EUR)
64,65
Online exclusive
71,60
Save 6,95 (10%)
Each
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
64,65
Online exclusive
71,60
Save 6,95 (10%)
Each
Chemical Identifiers
CAS7553-56-2
IUPAC Namediiodine
Molecular FormulaI2
InChI KeyPNDPGZBMCMUPRI-UHFFFAOYSA-N
SMILESII
View more
SpecificationsSpecification SheetSpecification Sheet
Insoluble matterPasses test
Appearance (Color)Dark grey
Appearance (Form)Metallic spheres
Titration Iodometric>=99.4 %
Chloride and Bromide=<0.005 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only