Copper(I) oxide, 97%
Copper(I) oxide, 97%
Copper(I) oxide, 97%
Thermo Scientific Chemicals

Copper(I) oxide, 97%

CAS: 1317-39-1 | Cu2O | 143.09 g/mol
Quantity:
25 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 197425000
Price (JPY)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1317-39-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Nickel (Ni)=<100 ppm
Silver (Ag)=<50 ppm
Titration Complexometric>=96.0 %
Iron (Fe)=<500 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only