Lead(II) iodide, 99%
Lead(II) iodide, 99%
Lead(II) iodide, 99%
Thermo Scientific Chemicals

Lead(II) iodide, 99%

CAS: 10101-63-0 | I2Pb | 461.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Glass bottle
Catalog number 198862500
Price (USD)
238.65
Online exclusive
265.00
Save 26.35
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
238.65
Online exclusive
265.00
Save 26.35
Each
Chemical Identifiers
CAS10101-63-0
IUPAC Nameλ²-lead(2+) diiodide
Molecular FormulaI2Pb
InChI KeyRQQRAHKHDFPBMC-UHFFFAOYSA-L
SMILES[I-].[I-].[Pb++]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=98.5 %
Chloride (Cl)=<100 ppm
Appearance (Color)Yellow to orange
Loss on drying=<0.1 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only