Acenaphthene, 99%
Acenaphthene, 99%
Acenaphthene, 99%
Thermo Scientific Chemicals

Acenaphthene, 99%

CAS: 83-32-9 | C12H10 | 154.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Plastic bottle
Catalog number 201345000
Price (USD)
96.65
Special offer
Online exclusive
Ends: 28-Dec-2026
114.00
Save 17.35
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
96.65
Special offer
Online exclusive
Ends: 28-Dec-2026
114.00
Save 17.35
Each
Chemical Identifiers
CAS83-32-9
IUPAC Name1,2-dihydroacenaphthylene
Molecular FormulaC12H10
InChI KeyCWRYPZZKDGJXCA-UHFFFAOYSA-N
SMILESC1CC2=C3C1=CC=CC3=CC=C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to brown
Infrared spectrumConforms
Melting point91°C to 95°C
Appearance (Form)Crystalline powder or crystals and/or chunks
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only