Amberlite™ XAD-7 HP
Amberlite™ XAD-7 HP
Amberlite™ XAD-7 HP
Thermo Scientific Chemicals

Amberlite™ XAD-7 HP

CAS: 37380-43-1
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 202241000
Price (EUR)
32,65
Online exclusive
36,20
Save 3,55 (10%)
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
32,65
Online exclusive
36,20
Save 3,55 (10%)
Each
Chemical Identifiers
CAS37380-43-1
MDL NumberMFCD00132705
SpecificationsSpecification SheetSpecification Sheet
TOC-AFNOR test=<10 ppm (total)
Particle size=<8.0 % (> 1.180 mm)
Uniformity coefficient=<2.0
Particle size=<7.0 % (< 0.300 mm)
Appearance (Color)White to pink-beige
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ion exchange resin
RUO – Research Use Only