Leuco Crystal Violet, 99%, pure
Leuco Crystal Violet, 99%, pure
Leuco Crystal Violet, 99%, pure
Thermo Scientific Chemicals

Leuco Crystal Violet, 99%, pure

CAS: 603-48-5 | C25H31N3 | 373.54 g/mol
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 204330250
Price (JPY)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS603-48-5
IUPAC Name4-{bis[4-(dimethylamino)phenyl]methyl}-N,N-dimethylaniline
Molecular FormulaC25H31N3
InChI KeyOAZWDJGLIYNYMU-UHFFFAOYSA-N
SMILESCN(C)C1=CC=C(C=C1)C(C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=98.5 %
Appearance (Form)Powder
Melting point175°C to 180°C
Appearance (Color)White to light grey to light violet
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only