ART™ Barrier Pipette Tips in Hinged Racks
ART™ Barrier Pipette Tips in Hinged Racks
ART™ Barrier Pipette Tips in Hinged Racks
Thermo Scientific™

ART™ Barrier Pipette Tips in Hinged Racks

Perform pipetting accurately and consistently with these barrier pipette tips in hinged racks, which seal when exposed to potential contaminants. ART hinged racks are available in low retention, micropoint, extended length and other specialty styles.
Catalog NumberVolume (Metric)ModelFormatTip Style
2069-HRPK200 μLART 200Hinged RackUniversal Fit
2139-HRPK10 μLART 10Hinged RackUniversal Fit
2139-05-HRPK10 μLART 10Hinged RackLow Retention
2149E-HRPK10 μLART 10 Ultra-microHinged RackUltra-micro
2149E-05-HRPK10 μLART 20E Ultra-microHinged RackLow Retention
2149P-HRPK20 μLART 20PHinged RackUniversal Fit
2149P-05-HRPK
also known as 2149P-05-HR
20 μLART 20PHinged RackLow Retention
2065-HRPK100 μLART 100Hinged RackUniversal Fit
2065E-HRPK100 μLART 100EHinged RackUniversal Fit
2065E-05-HRPK100 μLART 100EHinged RackLow Retention
2069-05-HRPK200 μLART 200Hinged RackLow Retention
2070-HRPK300 μLART BarrierRackUniversal Fit
2070-05-HRPK300 μLART BarrierRackLow Retention
2179-HRPK1000 μLART BarrierHinged RackUniversal Fit
Catalog number 2069-HRPK
Price (JPY)
19,600
Pack of 960
Volume (Metric):
200 μL
Model:
ART 200
Format:
Hinged Rack
Tip Style:
Universal Fit
Unlike standard filter tips, Thermo Scientific™ ART™ Barrier Hinged Rack Pipette Tips seal when exposed to potential contaminants. The pipette tips from the reload inserts fit securely into the environmentally friendly, reusable hinged racks and the covers may be used as hinged or lift-off style. The locking clasp prevents contamination by keeping remaining tips sterile and offers an easy, one-handed operation. ART hinged racks are available in low retention, micropoint, extended length, and other specialty styles.
Features:
  • Stackable hinged racks save space in your lab
  • Sterile tips are DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free

Recommended for:

  • Liquid Handling
  • Molecular Biology
  • PCR
  • Biotechnology
Specifications
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
DescriptionART 200, Filtered, Sterile, Hinged Rack
Length (English)2.32 in.
Length (Metric)5.88cm
ModelART 200
FilteredFiltered
Fit for PurposeSterile and PCR Ready
For Use With (Equipment)Universal Fit
FormatHinged Rack
Product LineART
SterilitySterile
Tip StyleUniversal Fit
Volume (Metric)200 μL
Unit SizePack of 960