Potassium oxalate monohydrate, 99+%, for analysis
Potassium oxalate monohydrate, 99+%, for analysis
Potassium oxalate monohydrate, 99+%, for analysis
Thermo Scientific Chemicals

Potassium oxalate monohydrate, 99+%, for analysis

CAS: 6487-48-5 | C2H2K2O5 | 184.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 207715000
Price (USD)
114.00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
114.00
Each
Chemical Identifiers
CAS6487-48-5
IUPAC Namedipotassium hydrate oxalate
Molecular FormulaC2H2K2O5
InChI KeyQCPTVXCMROGZOL-UHFFFAOYSA-L
SMILESO.[K+].[K+].[O-]C(=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Chloride (Cl)=<50 ppm
Iron (Fe)=<10 ppm
Appearance (Form)Crystals or flakes
Sulfate (SO4)=<100 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only