Potassium permanganate, 99+%, for analysis
Potassium permanganate, 99+%, for analysis
Potassium permanganate, 99+%, for analysis
Thermo Scientific Chemicals

Potassium permanganate, 99+%, for analysis

CAS: 7722-64-7 | KMnO4 | 158.032 g/mol
Quantity:
25 g
500 g
1 kg
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 207745000
Price (JPY)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7722-64-7
IUPAC Namepotassium manganesoylolate
Molecular FormulaKMnO4
InChI KeyVZJVWSHVAAUDKD-UHFFFAOYSA-N
SMILES[K+].[O-][Mn](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark green to purple to black
Appearance (Form)Glistening crystalline powder or crystals
Titration Iodometric99.0 to 101.0 %
Heavy metals (as Pb)=<30 ppm
Nitrogen compounds (as N)=<0.01 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only