Sodium bromate, 99+%, extra pure
Sodium bromate, 99+%, extra pure
Sodium bromate, 99+%, extra pure
Thermo Scientific Chemicals

Sodium bromate, 99+%, extra pure

CAS: 7789-38-0 | BrNaO3 | 150.9 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 208871000
Price (USD)
43.65
Online Exclusive
48.80
Save 5.15 (11%)
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
43.65
Online Exclusive
48.80
Save 5.15 (11%)
Each
Chemical Identifiers
CAS7789-38-0
IUPAC Namesodium bromate
Molecular FormulaBrNaO3
InChI KeyXUXNAKZDHHEHPC-UHFFFAOYSA-M
SMILES[Na+].[O-][Br](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfate (SO4)=<500 ppm
Appearance (Form)Crystalline powder
Heavy metals (as Pb)=<50 ppm
Bromide (Br)=<500 ppm
Titration Iodometric>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only