Specifications
CAS73-22-3
Chemical Name or MaterialL(-)-Tryptophan
ColorWhite
Melting Point290°C
Molecular FormulaC11H12N2O2
PackagingPoly bottle, tamper-evident cap
SynonymL-α-Amino-3-indolepropionic acid, (S)-2-Amino-3-(3-indolyl)propionic acid, H-Trp-OH, L-Tryptophan, Tryptophan
GradeMulti-Compendial/USP
Molecular Weight (g/mol)204.23
Percent Purity98.5 to 101.5%
Quantity1 kg
pH5.5 to 7
Unit SizeEach