(-)-Camphanic acid chloride, 98%
(-)-Camphanic acid chloride, 98%
(-)-Camphanic acid chloride, 98%
Thermo Scientific Chemicals

(-)-Camphanic acid chloride, 98%

CAS: 39637-74-6 | C10H13ClO3 | 216.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Plastic bottle
Catalog number 209890010
Price (USD)
-
Quantity:
1 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS39637-74-6
IUPAC Name(1S,4R)-4,7,7-trimethyl-3-oxo-2-oxabicyclo[2.2.1]heptane-1-carbonyl chloride
Molecular FormulaC10H13ClO3
InChI KeyPAXWODJTHKJQDZ-VHSXEESVSA-N
SMILESCC1(C)[C@@]2(C)CC[C@@]1(OC2=O)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals or flakes
Melting point67°C to 71°C
Specific optical rotation-17° to -25° (20°C, 589 nm) (c=3.5, CHCl3)
Titration RCOCl>=97.5 %
Appearance (Color)White to beige to grey
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chiral derivatization reagent
RUO – Research Use Only