Pierce™ Streptavidin, Maleimide-Activated
Thermo Scientific™
Pierce™ Streptavidin, Maleimide-Activated
Thermo Scientific Pierce Maleimide-Activated Streptavidin conjugate include streptavidin in a purified form, activated for crosslinking to sulfhydryl groups containing molecules.RelatedRead more
Have Questions?
Catalog NumberQuantity
211021 mg
Catalog number 21102
Price (USD)
269.00
Each
Quantity:
1 mg
Request bulk or custom format
Price (USD)
269.00
Each
Thermo Scientific Pierce Maleimide-Activated Streptavidin conjugate include streptavidin in a purified form, activated for crosslinking to sulfhydryl groups containing molecules.
Related Products
Pierce™ Streptavidin
Pierce™ Streptavidin, Horseradish Peroxidase Conjugated
Pierce™ Streptavidin, Alkaline Phosphatase Conjugated
Pierce™ Streptavidin, Hydrazide-Activated
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Reactivitythiol
DescriptionPierce Streptavidin, Maleimide Activated
FormulationLyophilized powder from protein in buffered saline solution
Label or DyeStreptavidin
Product TypeStreptavidin Conjugate
Quantity1 mg
Reactive MoietyMaleimide
SolubilitySoluble in Water
ConjugateStreptavidin
FormSolid
Product LinePierce
Unit SizeEach
Contents & Storage
Storage: Upon receipt store at room temperature or 4°C. Product shipped at ambient temperature.
Frequently asked questions (FAQs)
What is the molecular weight of Pierce Streptavidin, Maleimide-Activated (Cat. No. 21102)?
The molecular weight is 60 kDa.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.