Pierce™ Streptavidin, Hydrazide-Activated
Thermo Scientific™
Pierce™ Streptavidin, Hydrazide-Activated
Thermo Scientific Pierce Hydrazide-Activated Streptavidin conjugate include recombinant streptavidin in a purified form activated for crosslinking to carbonyl groups inRead more
Have Questions?
Catalog NumberQuantity
211202 mg
Catalog number 21120
Price (USD)
169.65
Online exclusive
226.00
Save 56.35
Each
Quantity:
2 mg
Request bulk or custom format
Price (USD)
169.65
Online exclusive
226.00
Save 56.35
Each
Thermo Scientific Pierce Hydrazide-Activated Streptavidin conjugate include recombinant streptavidin in a purified form activated for crosslinking to carbonyl groups in a molecule.
Related Products
Pierce™ Streptavidin
Pierce™ Streptavidin, Horseradish Peroxidase Conjugated
Pierce™ Streptavidin, Alkaline Phosphatase Conjugated
Pierce™ Streptavidin, Maleimide-Activated
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityKetone, Aldehyde
DescriptionPierce Streptavidin, Hydrazide Activated
Label or DyeStreptavidin
Product TypeStreptavidin Conjugate
Quantity2 mg
Reactive MoietyAmine, Hydrazide
SolubilitySoluble in Water
ConjugateStreptavidin
FormSolid
Product LinePierce
Unit SizeEach
Contents & Storage
Storage: Upon receipt store at room temperature or 4°C. Product shipped at ambient temperature.