Pierce™ Streptavidin
Thermo Scientific™
Pierce™ Streptavidin
Purified recombinant streptavidin enables detection of biotinylated antibodies and other probes in a variety of standard assay methods including western blotting, ELISA, immunohistochemistry, and fluorescence imaging.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
211221 mg
211255 mg
21135100 mg
211451000 mg
카탈로그 번호 21122
제품 가격(KRW)
232,000
온라인 행사
Ends: 31-Dec-2026
272,000
할인액 40,000
Each
수량:
1 mg
대량 주문 또는 맞춤형 요청
제품 가격(KRW)
232,000
온라인 행사
Ends: 31-Dec-2026
272,000
할인액 40,000
Each
Thermo Scientific Pierce Streptavidin is a purified unconjugated recombinant protein form.
Features of Streptavidin:
  • Recombinant streptavidin—high-quality protein, 53kDa, pI = 6.8 to 7.5
  • Pure protein —choose pure, unconjugated protein for biotin blocking and controls
The purified recombinant Streptavidin enables detection of biotinylated antibodies and other probes in a variety of standard assay methods, including Western blotting, ELISA, immunohistochemistry and fluorescence imaging. Purified streptavidin is supplied as lyophilized in phosphate-based buffers for immediate reconstitution with water.
Streptavidin is biotin-binding protein that was originally isolated from Streptomyces avidinii. In contrast to avidin, streptavidin has no carbohydrate and has a mildly acidic pI of 5. Thermo Scientific Pierce Streptavidin products use a recombinant form of streptavidin having a mass of 53,000 daltons and a near-neutral pI. Streptavidin is a tetrameric protein, with each subunit binding one molecule of biotin with affinity similar to that of avidin. Guanidinium chloride will dissociate avidin and streptavidin into subunits, but streptavidin is more resistant to dissociation.
Related Products
  • Pierce™ Streptavidin, Horseradish Peroxidase Conjugated (Cat. No. 21126)
  • Pierce™ Streptavidin, Alkaline Phosphatase Conjugated (Cat. No. 21324)
  • Pierce™ Streptavidin, Maleimide-Activated (Cat. No. 21102)
  • Pierce™ Streptavidin, Hydrazide-Activated (Cat. No. 21120)
For Research Use Only. Not for use in diagnostic procedures.
사양
라벨 또는 염료Unlabeled
제품 유형Streptavidin
수량1 mg
타겟 분자Biotin
콘주게이트Unconjugated
형태Solid
제품라인Pierce
Unit SizeEach
구성 및 보관
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C.
자주 묻는 질문(FAQ)
How many biotin binding-sites are there per streptavidin molecule?
Streptavidin is a protein composed of four identical subunits, each containing a high affinity binding site for biotin (K-D = 10 -15 M) . Streptavidin has the same biotin binding properties as avidin, but it has a low isoelectric point (pI=5) and no carbohydrate groups, resulting in low non-specific binding.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.