Trolox™, 97%
Trolox™, 97%
Trolox™, 97%
Thermo Scientific Chemicals

Trolox™, 97%

CAS: 53188-07-1 | C14H18O4 | 250.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 218940250
Price (USD)
663.65
Special offer
Online exclusive
Ends: 28-Dec-2026
781.00
Save 117.35
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
663.65
Special offer
Online exclusive
Ends: 28-Dec-2026
781.00
Save 117.35
Each
Chemical Identifiers
CAS53188-07-1
IUPAC Name6-hydroxy-2,5,7,8-tetramethyl-3,4-dihydro-2H-1-benzopyran-2-carboxylic acid
Molecular FormulaC14H18O4
InChI KeyGLEVLJDDWXEYCO-UHFFFAOYNA-N
SMILESCC1=C(C)C2=C(CCC(C)(O2)C(O)=O)C(C)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point187°C to 194°C
Appearance (Color)White to light beige
Infrared spectrumConforms
Titration with NaOH>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only