Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Description: The Mouse Hematopoietic Lineage FITC Cocktails contains FITC-conjugated antibodies that can be used to identify, enrich and/or deplete cells committed to the T, B, NK, myeloid and erythroid lineages based on the expression of cell surface antigens.
This cocktail contains the following antibodies:
Anti-Mouse CD3 (17A2) FITC
Anti-Mouse CD45R (B220) (RA3-6B2) FITC
Anti-Mouse CD11b (M1/70) FITC
Anti-Mouse TER-119 (TER-119) FITC
Anti-Mouse Ly-G6 (Gr-1) (RB6-8C5) FITC
Applications Reported: The Mouse Hematopoietic Lineage FITC Cocktail has been reported for use in flow cytometric analysis.
Applications Tested: The Mouse Hematopoietic Lineage FITC Cocktail has been pre-titrated and tested by flow cytometric analysis of C57Bl/6 bone marrow cells. This can be used at 20 µL per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
Hematopoietic Lineage antibodies can be used to identify, enrich and/or deplete cells committed to the T, B, NK, myeloid and erythroid lineages based on their expression of cell surface antigens.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: 17A2; Hematopoietic Lineage Cocktail; M1/70; RA3-6B2; RB6-8C5; TER-119
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support