Nalgene™ LDPE, Round Carboy
Thermo Scientific™

Nalgene™ LDPE, Round Carboy

For Use With: Store and transport reagents; Closure Material: Polypropylene; Color: White
Catalog NumberCapacity (Metric)
2210-0050PK20 L
2210-0040PK15 L
Catalog number 2210-0050PK
Price (JPY)
18,300
Each
Capacity (Metric):
20 L

Store and transport reagents with the Thermo Scientific™ Nalgene™ Round LDPE Carboys, featuring wide shoulder handles that allow for easy carrying and pouring with gloved hands. These carboys accept 13.5 rubber stoppers for an alternative sealing method.

  • Molded-in graduations of 5 L and 1 gal. increments
  • Leakproof†

Includes:White polypropylene screw cap and 83B closure

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
Capacity (English)5.3 gal.
Closure MaterialPolypropylene
Closure Size83B
Diameter (Metric) Inner Neck64 mm
Diameter (Metric) Outer286 mm
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Storage, Transportation ​
Height (Metric) With Closure528 mm
IncludesHandles
No. per Case4/Cs.
No. per Pack1 Ea.
ShapeRound
Workflow StepUpstream process solution storage. Upstream Validation
AutoclavableNot Autoclavable
Capacity (Metric)20 L
ColorWhite
MaterialLDPE
Product TypeCarboy with Handle
SterilityNon-sterile
Unit SizeEach