Dionex™ Inuvion™ Core IC System
Dionex™ Inuvion™ Core IC System
Dionex™ Inuvion™ Core IC System
Dionex™ Inuvion™ Core IC System
Thermo Scientific™
Dionex™ Inuvion™ Core IC System
Green features
Analyze ions with confidence using the versatile, affordable Thermo Scientific™ Dionex™ Inuvion™ Core Ion Chromatography System.
Catalog NumberMax. Pressure
22185-6010035 MPa (5000 psi)
Catalog number 22185-60100
Price (JPY)
-
Max. Pressure:
35 MPa (5000 psi)
Experience the simplicity of chemical suppression with faster, high-pressure operation. The Thermo Scientific™ Dionex™ Inuvion™ Core IC system makes ion analysis simpler and more intuitive, while providing consistent, high-resolution results with minimal training required. This system includes the robust features of the Dionex Inuvion platform while being easily configurable and offering affordable upgrades to full-featured capabilities when your analysis requirements increase.
  • The thermostatted high-performance conductivity detector permits measurements that are unaffected by temperature variation for improved reproducibility.
  • Advanced single-range digital output with operating range to 18,000 μS full scale, with autoranging to provide accurate detection of major and minor constituents in a single run. Single-range analog signal output is also standard.
  • Inert, nonmetallic, PEEK components throughout the system ensure compatibility with corrosive eluents and provide metal-contamination-free chromatography.
  • An electronically actuated six-port Rheodyne PEEK injection valve provides precise sampling.
Specifications
DescriptionDionex Inuvion Core IC System
Green FeaturesACT Ecolabel (Silver)
Max. Pressure35 MPa (5000 psi)
Detector TypeConductivity (CD)
Product TypeIC System
Unit SizeEach