Dionex™ Inuvion™ Auxiliary Valves
Thermo Scientific™
Dionex™ Inuvion™ Auxiliary Valves
Extend the application capability of your Thermo Scientific™ Dionex™ Inuvion™ IC system with auxiliary valves.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionPressure
22185-627042-Position Auxiliary Valve (6 port)35 MPa (5000 psi)
22185-627032-position Auxiliary Valve (10 port)35 MPa (5000 psi)
B51001290Auxiliary Valve0.7 MPa (100 psi)
Catalog number 22185-62704
Price (USD)
1,950.00
Each
Description:
2-Position Auxiliary Valve (6 port)
Pressure:
35 MPa (5000 psi)
Price (USD)
1,950.00
Each
Extend the application capability of your Thermo Scientific™ Dionex™ Inuvion™ IC system with auxiliary valves. The Dionex Inuvion IC system can support up to two additional low-pressure valves or one additional 6-port or 10-port high-pressure valve.
Optional accessory for all Dionex Inuvion IC system configurations
  • One 6-port high-pressure auxiliary valve
  • One 10-port high-pressure auxiliary valve
  • Two 3-port low-pressure auxiliary valves
Specifications
Description2-Position Auxiliary Valve (6 port)
For Use With (Equipment)Dionex Inuvion IC System
Pressure35 MPa (5000 psi)
TypeHigh-Pressure Valve
Unit SizeEach