Thermo Scientific Pierce BMPH is a short, maleimide-and-hydrazide crosslinker for conjugating sulfhydryls (cysteines) to carbonyls (aldehyde or ketones, such as자세히 알아보기
Have Questions?
카탈로그 번호
수량
22297
50 mg
카탈로그 번호 22297
제품 가격(KRW)
216,000
온라인 행사
Ends: 31-Dec-2026
253,000
할인액 37,000
Each
수량:
50 mg
대량 주문 또는 맞춤형 요청
제품 가격(KRW)
216,000
온라인 행사
Ends: 31-Dec-2026
253,000
할인액 37,000
Each
Thermo Scientific Pierce BMPH is a short, maleimide-and-hydrazide crosslinker for conjugating sulfhydryls (cysteines) to carbonyls (aldehyde or ketones, such as those formed by oxidation of glycoprotein carbohydrates).
Features of BMPH:
• Reactive groups:maleimide and hydrazide • Reactive towards: sulfhydryl groups and carbonyl (aldehyde) groups • Short (8.1A), sulfhydryl-to-aldehyde crosslinker with simple spacer arm (noncleavable) • Maleimide group reacts with sulfhydryl groups to form stable thioether linkages • Hydrazide group conjugates to oxidized sugars of glycoproteins and carbohydrates • Use sodium meta-periodate to oxidize glycosylation (e.g., sialic acid) to reactive aldehyde groups • Use with EDC to conjugate primary amine of hydrazide group to carboxyl groups
Product References:
Crosslinker Application Guide -- search for recent literature references for this product
For Research Use Only. Not for use in diagnostic procedures.
사양
세포 투과성Yes
설명BMPH
형태Solid
라벨링 방법Chemical Labeling
분자량297.19
페길레이티드No
제품라인Pierce
수량50 mg
반응성 부분Amine, Hydrazide, Maleimide
배송 조건Ambient
용해도Water
스페이서 암 길이8.1 Å
수용성Yes
화학물질 반응성Carbohydrate-Sulfhydryl
CleavableNo
교차결합제 유형Heterobifunctional
형식Standard
제품 유형Crosslinker
스페이서Short (<10 Å)
Unit SizeEach
구성 및 보관
Upon receipt store desiccated at 4°C.
인용 및 참조 문헌 (2)
인용 및 참조 문헌
Abstract
Protease domain glycans affect oligomerization, disulfide bond formation, and stability of the meprin A metalloprotease homo-oligomer.
Authors:Ishmael SS, Ishmael FT, Jones AD, Bond JS
Journal:J Biol Chem
PubMed ID:17040911
'The meprin A homo-oligomer is a highly glycosylated, secreted zinc metalloprotease of the astacin family and metzincin superfamily. This isoform of meprin is composed of disulfide-bonded dimers of alpha subunits that further associate to form large, secreted megadalton complexes of 10 or more subunits. The aim of this study was ... More
Interaction with the 5D3 monoclonal antibody is regulated by intramolecular rearrangements but not by covalent dimer formation of the human ABCG2 multidrug transporter.
Authors:Ozvegy-Laczka C, Laczkó R, Hegedus C, Litman T, Várady G, Goda K, Hegedus T, Dokholyan NV, Sorrentino BP, Váradi A, Sarkadi B
Journal:J Biol Chem
PubMed ID:18644784
Human ABCG2 is a plasma membrane glycoprotein working as a homodimer or homo-oligomer. The protein plays an important role in the protection/detoxification of various tissues and may also be responsible for the multidrug-resistant phenotype of cancer cells. In our previous study we found that the 5D3 monoclonal antibody shows a ... More