Tetrakis(triphenylphosphine)nickel(0), 95%
Tetrakis(triphenylphosphine)nickel(0), 95%
Tetrakis(triphenylphosphine)nickel(0), 95%
Thermo Scientific Chemicals

Tetrakis(triphenylphosphine)nickel(0), 95%

CAS: 15133-82-1 | C72H60NiP4 | 1107.86 g/mol
数量:
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 223980050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS15133-82-1
IUPAC Nametetrakis(triphenylphosphane) nickel
Molecular FormulaC72H60NiP4
InChI KeyKFBKRCXOTTUAFS-UHFFFAOYSA-N
SMILES[Ni].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
さらに表示
Appearance (Color)Rust brown to brown
Appearance (Form)Powder or crystals
Elemental Analysis>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only