Nalgene™ Autoclavable Polycarbonate Lowboys with Spigot, 15L - FAQs
Nalgene™ Autoclavable Polycarbonate Lowboys with Spigot, 15L - FAQs
View additional product information for Nalgene™ Autoclavable Polypropylene Copolymer (PPCO) Lowboys with Spigot - FAQs (2324-0015)
1 product FAQs found
Do you offer replacement carboy spigots?
The standard Nalgene polyproylene quick action spigot (blue handle and body, white threaded ring for connection to carboy) Cat. No. is 6422-0010 or 6422-0010PK. If the original carboy purchased was a solvent-resistant fluorinated carboy, the replacement (mostly white, with a blue cap on the handle) is the Nalgene Quick-Action Tefzel ETFE Spigot, Cat. No. 6432-0010.