Methyl trifluoroacetate, 99%
Methyl trifluoroacetate, 99%
Methyl trifluoroacetate, 99%
Thermo Scientific Chemicals

Methyl trifluoroacetate, 99%

CAS: 431-47-0 | C3H3F3O2 | 128.05 g/mol
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 243411000
Price (JPY)
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS431-47-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only