2-Mercaptopyridine-N-oxide, sodium salt, 40 w/w % aqueous solution
2-Mercaptopyridine-N-oxide, sodium salt, 40 w/w % aqueous solution
2-Mercaptopyridine-N-oxide, sodium salt, 40 w/w % aqueous solution
Thermo Scientific Chemicals

2-Mercaptopyridine-N-oxide, sodium salt, 40 w/w % aqueous solution

CAS: 3811-73-2 | C5H4NNaOS | 149.15 g/mol
Quantity:
1 L
2.5 L
5 L
Packaging:
View all
Glass bottle
Catalog number 257740010
Price (JPY)
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3811-73-2
IUPAC Namesodium 2-sulfanylidene-1,2-dihydropyridin-1-olate
Molecular FormulaC5H4NNaOS
InChI KeyXNRNJIIJLOFJEK-UHFFFAOYSA-N
SMILES[Na+].[O-]N1C=CC=CC1=S
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO440 to 42 %
Appearance (Color)Yellow to amber
pH8.5 to 10.5 (10 % at 20°C)
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only