Pierce™ Anti-HA Agarose
Thermo Scientific™

Pierce™ Anti-HA Agarose

Thermo Scientific Pierce Anti-HA Agarose is an immunopurification and immunoprecipitation resin for the enrichment of HA-tagged proteins expressed in human자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
261811 mL
261825 mL
카탈로그 번호 26181
제품 가격(KRW)
925,000
온라인 행사
Ends: 31-Dec-2026
1,027,000
할인액 102,000 (10%)
Each
수량:
1 mL
대량 주문 또는 맞춤형 요청
제품 가격(KRW)
925,000
온라인 행사
Ends: 31-Dec-2026
1,027,000
할인액 102,000 (10%)
Each

Thermo Scientific Pierce Anti-HA Agarose is an immunopurification and immunoprecipitation resin for the enrichment of HA-tagged proteins expressed in human in vitro protein expression systems, bacteria, yeast, and mammalian cells. The anti-HA antibody coupled to the resin is a high-affinity mouse IgG1 monoclonal antibody that recognizes the HA-epitope tag (YPYDVPDYA) derived from the human influenza hemagglutinin (HA) protein.

Features of Pierce Anti-HA Agarose

  • Specific—highly specific anti-HA monoclonal antibody enables high yield and high purity
  • Scalable—available in 1 and 5 mL settled resin package sizes to allow for smaller scale immunoprecipitations or larger scale purifications
  • Versatile—can be used in spin or gravity-based purification and enrichment methods

Pierce Anti-HA Agarose consists of highly specific monoclonal anti-HA antibody that is covalently immobilized on a crosslinked 4% beaded agarose support. The product is supplied as a 50% slurry in 0.1 M phosphate, 0.15 M NaCl, pH 7.2 with 0.05% sodium azide. The anti-HA agarose has 3.5 mg mouse anti-HA IgG1 monoclonal antibody conjugated per mL of settled agarose resin.

HA-tagged proteins are easily eluted from the resin using 0.1 M glycine (pH 2.0–2.8), 3 M NaSCN, or 50 mM NaOH, depending on the downstream application of the purified protein. For gentle elution, HA Synthetic Peptide is available for use in neutralization and to competitively elute HA-tagged fusion proteins from immobilized anti-HA affinity resin.

Related products

Pierce HA-Tag IP/Co-IP Kit

Pierce HA-Tag Magnetic IP/Co-IP Kit

For Research Use Only. Not for use in diagnostic procedures.
사양
콘주게이트HA-epitope tag (YPYDVPDYA)
정제 타겟Anti-HA Antibody
수량1 mL
컬럼 유형Agarose Resin, Affinity
형식Bottle
제품 유형Agarose
Unit SizeEach
구성 및 보관
1 mL of settle resin, provided as 2 mL of 50% slurry. Store at 2–8°C.

자주 묻는 질문(FAQ)

What type of recombinant protein purification resins do you offer?

We offer a variety of resins targeting His-tagged fusion proteins or GST fusion proteins. Please see this selection table to choose the right resin for your experiments. Table available at http://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-purification-isolation/protein-purification/recombinant-protein-purification.html

We also offer resins (both agarose and magnetic bead-based) with immobilized anti-c-Myc and anti-HA antibodies for targeting c-Myc and HA-tagged proteins respectively. Please check out the following links for the specific products:

-Anti-c-Myc agarose (https://www.thermofisher.com/order/catalog/product/20169?ICID=search-product)
-Anti-c Myc magnetic beads (https://www.thermofisher.com/order/catalog/product/88843?ICID=search-product)
-Anti-HA agarose (https://www.thermofisher.com/order/genome-database/antibody/HA-Epitope-Tag-Antibody-clone-2-2-2-14-Monoclonal/26181)
-Anti-HA magnetic beads: (https://www.thermofisher.com/order/catalog/product/88837?ICID=search-product)


Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.