trans-2-Amino-1-cyclohexanecarboxylic acid, 98%
trans-2-Amino-1-cyclohexanecarboxylic acid, 98%
trans-2-Amino-1-cyclohexanecarboxylic acid, 98%
Thermo Scientific Chemicals

trans-2-Amino-1-cyclohexanecarboxylic acid, 98%

CAS: 5691-19-0 | C7H13NO2 | 143.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Catalog number 265630050
Price (TWD)
-
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS5691-19-0
IUPAC Name(1R,2R)-2-azaniumylcyclohexane-1-carboxylate
Molecular FormulaC7H13NO2
InChI KeyUSQHEVWOPJDAAX-PHDIDXHHSA-N
SMILES[NH3+][C@@H]1CCCC[C@H]1C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder and/or chunks
Titration with HClO4>=97.5 %
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only