2'-OMe-PAC-A Phosphoramidite, standard grade, 20-400 finish bottle
Thermo Scientific™

2'-OMe-PAC-A Phosphoramidite, standard grade, 20-400 finish bottle

2'-OMe-PAC-A CEP is a phosphoramidite containing 5' DMT and the nucleoside adenosine (A) with 2'-O-methyl protection, for use in oligonucleotideRead more
Catalog NumberQuantity
27-1822-101 g
Catalog number 27-1822-10
Price (JPY)
-
Quantity:
1 g

2'-OMe-PAC-A CEP is a phosphoramidite containing 5' DMT and the nucleoside adenosine (A) with 2'-O-methyl protection, for use in oligonucleotide synthesis. Located in the ideal oligo position, 2'-O-methyl (OMe) oligo modifications display nuclease resistance, fewer off-target effects, and slightly increased hybridization affinities. These phosphoramidites are commonly used for oligos designed for antisense, siRNA, aptamer, or gRNA-based research. The 'standard grade' marking denotes a baseline level of purity verified with analytical methods, and proven to be suitable for most oligo synthesis requirements. This product is supplied in a 30-mL amber bottle with a 20-400 finish neck style, which is compatible with most Expedite synthesizers.

Features:
• Full chemical name—5'-O-(4,4'-Dimethoxytrityl)-N6-phenoxyacetyl-2'-O-methyl-adenosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula—C49H56N7O9P
• Base—A
• Base Protection—PAC
• Formula weight—918.0 g/mol
• CAS number—128219-82-9
• HPLC purity—≥98.0%
• 31P NMR purity—≥98.0%

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Expedite Synthesizers
FormPowder
GradeStandard
Nucleic Acid TypeRNA
Nucleoside TypeAdenosine (A)
Product Type2in-OMe-PAC-A CEP
Purity98%
Quantity1 g
Reagent Features2in-OMe modified, Fast deprotection
Reagent TypePhosphoramidite
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store in freezer (at or below -20°C).