(-)-Terpinen-4-ol, 97%, sum of enantiomers
(-)-Terpinen-4-ol, 97%, sum of enantiomers
(-)-Terpinen-4-ol, 97%, sum of enantiomers
Thermo Scientific Chemicals

(-)-Terpinen-4-ol, 97%, sum of enantiomers

CAS: 20126-76-5 | C10H18O | 154.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 270430250
Price (USD)
72.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
72.00
Each
Chemical Identifiers
CAS20126-76-5
IUPAC Name(1R)-4-methyl-1-(propan-2-yl)cyclohex-3-en-1-ol
Molecular FormulaC10H18O
InChI KeyWRYLYDPHFGVWKC-JTQLQIEISA-N
SMILESCC(C)[C@@]1(O)CCC(C)=CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation-19° to -31° (20°C, 589 nm) (neat)
Infrared spectrumConforms
GC>=96.0 % (sum of enantiomers)
Density(20/4°C) 0.930 to 0.936 g/ml
Refractive index1.4760 to 1.4820 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only