9-Hydroxyxanthene, 97+%
9-Hydroxyxanthene, 97+%
9-Hydroxyxanthene, 97+%
9-Hydroxyxanthene, 97+%
Thermo Scientific Chemicals

9-Hydroxyxanthene, 97+%

CAS: 90-46-0 | C13H10O2 | 198.22 g/mol
Quantity:
5 g
25 g
Packaging:
View all
Plastic bottle
Catalog number 271520250
Price (JPY)
-
Request A Quote
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS90-46-0
IUPAC Name9H-xanthen-9-ol
Molecular FormulaC13H10O2
InChI KeyJFRMYMMIJXLMBB-UHFFFAOYSA-N
SMILESOC1C2=CC=CC=C2OC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
HPLC>=97.0 %
Infrared spectrumConforms
Melting point121°C to 126°C
Appearance (Color)White to almost white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only