4-Methoxybenzylchloride, 98%, stabilized
4-Methoxybenzylchloride, 98%, stabilized
4-Methoxybenzylchloride, 98%, stabilized
Thermo Scientific Chemicals

4-Methoxybenzylchloride, 98%, stabilized

CAS: 824-94-2 | C8H9ClO | 156.61 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
100 g
產品號碼 294171000
價格 (TWD)
-
Quantity:
100 g
Request bulk or custom format
規格
Assay Percent Range97.5% min. (GC)
CAS Max %100.0
CAS Min %97.5
CAS824-94-2
CAS584-08-7
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only