Nalgene™ Reusable Filter Units
Thermo Scientific™
Nalgene™ Reusable Filter Units
Eliminate the need for a manifold or breakable glass vacuum flask. Nalgene Reusable Filter Holders with Receiver are complete, environmentally-friendly systems.
Catalog NumberVolume (Metric) Receiver
300-4000PK
also known as 300-4000
250 mL
300-4050PK500 mL
300-4100PK1000 mL
Catalog number 300-4000PK
also known as 300-4000
Price (JPY)
24,820
Each
Volume (Metric) Receiver:
250 mL
Reusable filter units include two membrane support plates: a clear one-piece plate for sterilization applications and a two-piece white plate for analytical applications. Accepts 47 or 50 mm diameter membranes. Two side-arms allow connection to vacuum line. Tubing adapters accept 1/4- to 5/16-inch (6 to 8 mm) I.D. vacuum tubing.
Features
  • Replaces metal or breakable glass – major components are made of durable, transparent, autoclavable polysulfone
  • Use for sterilization or analysis. Filter holders come with sterilization and analytical membrane support plates
  • Designed to be used with vacuum or pressure
  • Graduated chambers for easy reference
Specifications
AutoclavableAutoclavable
No. per Pack1
Volume (Metric) Receiver250 mL
Volume (Metric) Upper250 mL
MaterialPolysulfone
Unit SizeEach