Rubidium nitrate, 99.8%, pure
Rubidium nitrate, 99.8%, pure
Rubidium nitrate, 99.8%, pure
Thermo Scientific Chemicals

Rubidium nitrate, 99.8%, pure

CAS: 13126-12-0 | NO3Rb | 147.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 300070500
Price (USD)
243.00
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
243.00
Each
Chemical Identifiers
CAS13126-12-0
IUPAC Namerubidium(1+) nitrate
Molecular FormulaNO3Rb
InChI KeyRTHYXYOJKHGZJT-UHFFFAOYSA-N
SMILES[Rb+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total trace metal impurities=<0.2 %
Appearance (Color)White
Appearance (Form)Crystals or needles
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only