Hypersil™ MOS-2 C8 HPLC Columns
Thermo Scientific™

Hypersil™ MOS-2 C8 HPLC Columns

Use as a general-purpose C8 phase for acidic and neutral analytes, with decrease silanol activity
Catalog NumberLength (Metric)Diameter (Metric)Particle Size
30305-154630150 mm4.6 mm5 μm
30305-254630250 mm4.6 mm5 μm
30305-124630125 mm4.6 mm3 μm
30305-01400110 mm4 mm5 μm
Catalog number 30305-154630
Price (JPY)
73,000
Each
Length (Metric):
150 mm
Diameter (Metric):
4.6 mm
Particle Size:
5 μm
For reliable, affordable analysis of more hydrophobic analytes such as small peptides and proteins, pharmaceuticals, steroids, and environmental samples, use Thermo Scientific™ Hypersil™ MOS-2 C8 HPLC Columns.

Hypersil stationary phases are recognized as an HPLC industry standard. A proven effective analytical tool, classical Hypersil phases are well established and referenced in HPLC methods. Quality is assured by manufacture under ISO 9001-2000 guidelines.

For separations requiring improved chromatographic performance, use Thermo Scientific™ Hypersil GOLD™ C8 columns as an alternative.

  • Reliable, less retentive phase
  • Long column lifetimes, even under basic conditions
  • Manufactured under ISO 9001-2000 quality guidelines
  • Affordable
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column
Column TypeReversed Phase
Max. Pressure5800 psi (400 bar)
Packing MaterialSpherical, Fully Porous Type A Silica
Stationary PhaseC8
Surface Area170 m²/g
Temperature60°C
USP TypeL7
Carbon Load6.5%
Diameter (Metric)4.6 mm
Length (Metric)150 mm
Particle Size5 μm
PhaseReversed Phase
Pore Size120 Å
Product LineHypersil Classical
pH2 to 8
Unit SizeEach