Sarcosine ethyl ester hydrochloride, 97%
Sarcosine ethyl ester hydrochloride, 97%
Sarcosine ethyl ester hydrochloride, 97%
Thermo Scientific Chemicals

Sarcosine ethyl ester hydrochloride, 97%

CAS: 52605-49-9 | C5H12ClNO2 | 153.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 305590100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS52605-49-9
IUPAC Namehydrogen ethyl 2-(methylamino)acetate chloride
Molecular FormulaC5H12ClNO2
InChI KeyNIDZUMSLERGAON-UHFFFAOYSA-N
SMILES[H+].[Cl-].CCOC(=O)CNC
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=96.0 %
Loss on drying=<1 %
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only