Calcium iodate, pure
Calcium iodate, pure
Calcium iodate, pure
Thermo Scientific Chemicals

Calcium iodate, pure

CAS: 7789-80-2 | CaI2O6 | 389.89 g/mol
Have Questions?
Catalog NumberQuantityPackaging
318411000100 gPlastic bottle
Catalog number 318411000
Price (USD)
39.65
Special offer
Online exclusive
Ends: 30-Jun-2026
46.80
Save 7.15 (15%)
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
39.65
Special offer
Online exclusive
Ends: 30-Jun-2026
46.80
Save 7.15 (15%)
Each
Chemical Identifiers
CAS7789-80-2
IUPAC Namecalcium diiodate
Molecular FormulaCaI2O6
InChI KeyUHWJJLGTKIWIJO-UHFFFAOYSA-L
SMILES[Ca++].[O-][I](=O)=O.[O-][I](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Appearance (Form)Crystalline powder
Titration Iodometric>=96 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only