9-Methyladenine, 98%, Thermo Scientific Chemicals
9-Methyladenine, 98%, Thermo Scientific Chemicals
9-Methyladenine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

9-Methyladenine, 98%, Thermo Scientific Chemicals

CAS: 700-00-5 | Molecular Formula: C6H7N5 | Molecular Weight: 149.16 g/mol
Have Questions?
Catalog NumberQuantityPackaging
325871000100 mgGlass bottle
Catalog number 325871000
Price (USD)
-
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS700-00-5
IUPAC Name9-methyl-9H-purin-6-amine
Molecular FormulaC6H7N5
InChI KeyWRXCXOUDSPTXNX-UHFFFAOYSA-N
SMILESCN1C=NC2=C(N)N=CN=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige
Appearance (Form)Crystals
HPLC>=97.5 %
Infrared spectrumConforms

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only