Cyclohexane, for pesticide residue analysis
Cyclohexane, for pesticide residue analysis
Cyclohexane, for pesticide residue analysis
Thermo Scientific Chemicals

Cyclohexane, for pesticide residue analysis

CAS: 110-82-7 | C6H12 | 84.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number 326590010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Beilstein05, 20
Boiling Point81.0°C
CAS110-82-7
ChEBICHEBI:29005
Chemical Name or MaterialCyclohexane
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For residue analysis
RUO – Research Use Only