Thermo Scientific Chemicals

TAPSO, 99+%, for biochemistry

Catalog number: 327811000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

TAPSO, 99+%, for biochemistry

Catalog number: 327811000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 327811000
Price (USD)
Price: 103.00
Online price: 87.65
Your price:
Quantity
-

Chemical Identifiers

CAS
68399-81-5
IUPAC Name
3-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}-2-hydroxypropane-1-sulfonic acid
Molecular Formula
C7H17NO7S
InChI Key
RZQXOGQSPBYUKH-UHFFFAOYNA-N
SMILES
OCC(CO)(CO)NCC(O)CS(O)(=O)=O
Appearance (Color)
White
Appearance (Form)
Powder
Titration with NaOH
>=99 %
Water
=<1 % (K.F.)
pH
4.3 to 5.3 (1 % in water at 25°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text