2-Butanone, 99+%, for electronic use (MOS), residue free
2-Butanone, 99+%, for electronic use (MOS), residue free
2-Butanone, 99+%, for electronic use (MOS), residue free
Thermo Scientific Chemicals

2-Butanone, 99+%, for electronic use (MOS), residue free

CAS: 78-93-3 | C4H8O | 72.11 g/mol
数量:
1 L
2.5 L
パッケージ:
Glass Bottle
製品番号(カタログ番号) 327910025
価格(JPY)
-
数量:
2.5 L
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS78-93-3
IUPAC Namebutan-2-one
Molecular FormulaC4H8O
InChI KeyZWEHNKRNPOVVGH-UHFFFAOYSA-N
SMILESCCC(C)=O
さらに表示
Appearance (Form)Clear liquid
GC>=99.0 %
Arsenic (As)=<20 ppb
Heavy metals (as Pb)=<1 ppm
Copper (Cu)=<10 ppb
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only