3-Butyn-2-one, 96%
3-Butyn-2-one, 96%
3-Butyn-2-one, 96%
Thermo Scientific Chemicals

3-Butyn-2-one, 96%

CAS: 1423-60-5 | C4H4O | 68.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
Catalog number 330440025
Price (USD)
-
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1423-60-5
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4045 to 1.4095 (20°C, 589 nm)
Appearance (Color)Clear yellow to orange-brown
GC>=95 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only